Learn More
Aat Bioquest XFD594 amine *Same Structure to Alexa Fluor™ 594 amine* 1mg
SDP
Supplier:  Aat Bioquest 1715
Encompass_Preferred
XFD594 amine is the same molecule to Alexa Fluor® 594 C5 amine (Alexa Fluor® is the trademark of ThermoFisher). The fluorescent dye amines are reactive molecules that can be used to add a fluorescent label to biomolecules containing a carbonyl group. XFD594 amine reacts with carboxylic acids, aldehydes and ketones. It is a very useful building block for developing XFD594 probes.
Catalog No. NC2826972