Learn More
Nanocs Alexa fluor 647 PEG Biotin, 5 mg
Supplier:  Nanocs PG2AF647BN3K
Encompass
Alexa fluor 647 labeled PEG Biotin (AF647 PEG Biotin is a near IR fluorescent PE biotin derivative having excitation/emission wavelength around 651 nm/672 nm. Alexa fluor 647 is a near infrared fluorescent dye with high fluorescent intensity and excellent photo stability. Alexa fluor 647 PEG biotin offered by Nanocs is a water soluble fluorescent biotin PEG that can be used directly in aqueous buffer without need adding toxic organic solvents.
Catalog No. NC3348086