Learn More
Biolegend Alexa Fluor 700 anti-mouse TCR Vα2
Supplier:  Biolegend 127824
Encompass_Preferred
Alexa Fluor 700 anti-mouse TCR Vα2 [B20.1]; Isotype: Rat IgG2a, λ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1766734