Learn More
Biolegend Alexa FluorR 488 anti-NRF2
Supplier:  Biolegend 939207
Encompass_Preferred
Alexa Fluor 488 anti-NRF2 [W19086B]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests
Catalog No. 50-286-6011