Learn More
TARGETMOL CHEMICALS INC ALFACALCIDOL 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T637425MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Alfacalcidol (Etalpha) (1-hydroxycholecalciferol; Alpha D3; 1.alpha.-Hydroxyvitamin D3) is a non-selective VDR activator medication. purity: 99%
Catalog No. 50-257-9406