Learn More
Cayman Chemical AlflutInIbmesylate 10mg
SDP
Supplier:  Cayman Chemical 3147310 MG
Encompass_Preferred
An inhibitor of mutant EGFRs; selective for EGFRL858R, EGFRG719X, EGFRL858R, EGFRL861A, and EGFRT790M mutant EGFRs over wild-type EGFR; reduces tumor growth in an EGFRL858R and EGFRT790M-expressing LU1868 NSCLC PDX mouse model at 10 and 30 mg/kg
Catalog No. 50-299-0816