Learn More
Selleck Chemical LLC Alfuzosin HCl S1409-10mg
SDP
Supplier:  Selleck Chemical LLC S140910MG
Encompass_Preferred
Alfuzosin HCI(SL 77499 HCl) is an alpha1 receptor antagonist used to treat benign prostatic hyperplasia (BPH)
Catalog No. 50-311-0708