Learn More
Medchemexpress LLC Alginate lyase | 9024-15-1 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP283510MG
Encompass_Preferred
Alginate lyase is a polysaccharide lyase that catalyzes the degradation of alginate. It can be utilized in research related to cystic fibrosis by degrading the polysaccharide biofilm of *Pseudomonas aeruginosa*.
  • Catalyzes the degradation of alginate.
  • Useful in cystic fibrosis research for degrading *Pseudomonas aeruginosa* biofilm.
  • Available in various quantities.
  • Enzyme activity: ≥10000 U/g solid.
  • Multiple batches available with associated documentation.
Catalog No. 50-003-99864