Learn More
Medchemexpress LLC Alginate lyase | 9024-15-1 | ≥10000 U/g | 5 MG

Supplier: Medchemexpress LLC HYP28355MG
Alginate lyase is a polysaccharide lyase that catalyzes the degradation of alginate. It can be used for the research of cystic fibrosis by degrading the polysaccharide biofilm of *Pseudomonas aeruginosa*.
- Catalyzes the degradation of alginate
- Used for the research of cystic fibrosis
- Degrades polysaccharide biofilm of *Pseudomonas aeruginosa*
- Appearance: Solid
- Color: Light yellow to light brown
- Solubility: H2O : ≥ 2.5 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.