Learn More
Medchemexpress LLC Alginate methacryloyl | 50,000 (average) | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15822210MG
Encompass_Preferred
Alginate methacryloyl (AlgMA) is a methacryloyl-functionalized alginate polymer that forms photocrosslinkable hydrogels for tissue engineering and biological 3D printing. It is biocompatible, non-immunogenic, and can physically cross-link with divalent cations; when combined with a photoinitiator it forms stable, cell-supportive hydrogel networks suitable for cell culture and scaffold fabrication.
  • Methacryloyl-functionalized alginate for photocrosslinkable hydrogel formation.
  • Average molecular weight ~50,000 Da.
  • Biocompatible and non-immunogenic.
  • Low intrinsic toxicity suitable for cell culture.
  • Physical cross-linking with divalent cations (e.g., calcium) and photoinitiated covalent cross-linking.
  • Suitable for biological 3D printing and tissue engineering scaffolds.
  • Supplied in laboratory-scale mg quantities.
Catalog No. 50-002-88383