Learn More
Medchemexpress LLC Alginic acid | 9005-32-7 | 81309 | 97.0% | (C6H8O6)n | 100 G
SDP
Supplier:  Medchemexpress LLC HYW127758100G
Encompass_Preferred
Alginic acid is a natural polysaccharide extracted from brown seaweeds, used in research for its film-forming, water-soluble, biodegradable, and biocompatible properties. It has reported bioactivities including inhibition of histamine release and effects on oxidative stress-mediated hormone secretion, apoptosis, and autophagy in mouse granulosa cells and ovaries.
  • Natural polysaccharide with good water solubility.
  • Forms films and hydrogels for formulation and material studies.
  • Biodegradable and biocompatible for biological applications.
  • Demonstrated inhibitory effect on histamine release.
  • Relevant for studies of oxidative stress, apoptosis, and autophagy.
  • Supplied as a dry powder suitable for laboratory handling.
Catalog No. 50-002-83624