Learn More
Medchemexpress LLC Alginic acid, calcium salt | 9005-35-0 | MFCD00143567 | (C12H14CaO12)n | 50 G
SDP
Supplier:  Medchemexpress LLC HYW13396750G
Encompass_Preferred
Calcium alginate (alginic acid, calcium salt) is a natural, pH-sensitive biopolymer used to form hydrogels for research applications such as oral agent-delivery systems, wound dressings, and gel-forming matrices. Supplied as a white to light-yellow solid, it is characterized by batch-level properties including moisture, ash content, viscosity, mesh size, and pH.
  • Biopolymer pH-sensitive hydrogel suitable for controlled release.
  • Stable in acidic media and soluble in basic media.
  • Forms biodegradable gel matrices for encapsulation and bead formation.
  • Provided as a dry solid for convenient handling and storage.
  • Batch-level specifications include moisture, ash, viscosity, mesh, and pH.
  • Intended for research use only; not for clinical or human use.
Catalog No. 50-002-83123