Learn More
Medchemexpress LLC Alginic acid, calcium salt | 9005-35-0 | MFCD00143567 | Varies (polymeric); reported molar masses 584.45 g/mol and 1,170.93 g/mol | Empirical: C18H24CaO19; polymeric repeat unit: (C12H14CaO12)n | 500 G
SDP
Supplier:  Medchemexpress LLC HYW133967500G
Encompass_Preferred
Calcium alginate is a biodegradable, pH-sensitive hydrogel (alginic acid, calcium salt) used as a gel-transforming agent and excipient. It is stable in acidic media and soluble in basic media, enabling pH-triggered absorption and release of aqueous solutions. Common applications include oral agent delivery, nasal packing, cell culture scaffolds, and tissue engineering.
  • Biodegradable pH-responsive hydrogel suitable for controlled release.
  • Stable in acidic environments and soluble in basic environments.
  • Enables pH-triggered absorption and release of aqueous solutions.
  • Useful for oral drug delivery systems and nasal packing applications.
  • Compatible with cell culture scaffolds and 3D bioprinting.
  • Provides a biodegradable matrix for tissue engineering and wound care.
Catalog No. 50-002-83504