Learn More
Research Products International Corp Alginic Acid Sodium Salt, 500 Grams
SDP
Supplier:  Research Products International Corp A50300500.0
Encompass_Preferred
From Phaeophyceae (algae). A mixture of polyuronic acids composed of residues of D-mannuronic acid and L-glucuronic acids. Alginates are used as suspending and thickening agents and in the preparation of water miscible gels. Viscosity: 1%, 25 cps.
Catalog No. 50-213-157