Learn More
Medchemexpress LLC Alimemazine hemitartrate | 4330-99-8 | 98.4% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12752A10MM/1ML
Encompass_Preferred
Alimemazine hemitartrate is an orally active antagonist of the histamine (HA) receptor and a partial agonist at other GPCR receptors. It exhibits various properties including anti-serotonin, antispasmodic, antihistamine, sedative, hypnotic, antiviral, and anti-nausea effects. It also promotes pancreatic β-cell growth and inhibits SARS-CoV-2.
  • Antagonist of histamine (HA) receptor.
  • Partial agonist at other GPCR receptors.
  • Exhibits anti-serotonin, antispasmodic, antihistamine, sedative, hypnotic, antiviral, and anti-nausea properties.
  • Promotes pancreatic β-cell growth and improves diabetes in mice.
  • Reduces the formation of syncytia induced by SARS-CoV-2 and its variants.
  • Inhibits SARS-CoV-2 in HEK293T/ACE2 and VeroE6/TMPRSS2 cells.
Catalog No. 50-003-41371