Learn More
Medchemexpress LLC Alimemazine hemitartrate | 4330-99-8 | 98.4% | 200 MG
SDP
Supplier:  Medchemexpress LLC HY12752A200MG
Encompass_Preferred
Alimemazine hemitartrate is an antagonist of the histamine (HA) receptor that also acts as a partial agonist at other GPCR receptors. This compound exhibits anti-serotonin, antispasmodic, antihistamine, sedation, hypnotic, antiviral, and anti-nausea properties. It promotes pancreatic β-cell growth and improves diabetes in mice. This product is intended for research use only.
  • Acts as a histamine (HA) receptor antagonist
  • Functions as a partial agonist at GPCR receptors
  • Exhibits anti-serotonin, antispasmodic, and antihistamine properties
  • Offers sedative, hypnotic, antiviral, and anti-nausea effects
  • Promotes pancreatic β-cell growth
  • Improves diabetes in mouse models
  • Reduces SARS-CoV-2 induced syncytia formation
  • Inhibits SARS-CoV-2 in HEK293T/ACE2 and VeroE6/TMPRSS2 cells
  • For research use only
Catalog No. 50-003-42664