Learn More
Medchemexpress LLC Alinidine hydrobromide | 71306-36-0 | 99.0% | 351.07 | C12H14BrCl2N3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY106688A1MG
Encompass_Preferred
Alinidine hydrobromide is the hydrobromide salt of alinidine, a research antiarrhythmic compound. It reduces the slope of diastolic depolarization in sinoatrial tissue and Purkinje fibers and has demonstrated anti-ischemic and antiarrhythmic effects. Intended for laboratory research use.
  • Antiarrhythmic and bradycardic activity demonstrated in cardiac tissue
  • Reduces slope of diastolic depolarization in sinoatrial tissue and Purkinje fibers
  • High purity (99.0%) suitable for research applications
  • Molecular weight 351.07 and formula C12H14BrCl2N3
  • Available in small-scale quantities for laboratory experiments (1 mg-100 mg)
  • For research use only; not for human or veterinary use
Catalog No. 50-002-42119