Learn More
Selleck Chemical LLC Alisertib (MLN8237) S1133-200mg
SDP
Supplier:  Selleck Chemical LLC S1133200MG
Encompass_Preferred
Alisertib (MLN8237) is a selective Aurora A inhibitor with IC50 of 1 2 nM in a cell-free assay It has 200-fold higher selectivity for Aurora A than Aurora B Alisertib induces cell cycle arrest apoptosis and autophagy Phase 3
Catalog No. 50-310-8501