Learn More
Apexbio Technology LLC Alisol A 19885-10-0 20mg
SDP
Supplier:  Apexbio Technology LLC N202120
Encompass_Preferred
Alisol A is a natural triterpenoid compound isolated from the tubers of Alisma plantago-aquatica L var orientale Samuels (CAS 19885-10-0) Preclinical studies indicate that Alisol A modulates lipid metabolism and exhibits anti-inflammatory properties by regulating key signaling pathways involved in metabolic and inflammatory responses The compound is under active investigation for its potential to influence metabolic disorders inflammation and related cellular mechanisms Alisol A serves as a valuable molecular probe in pharmacological research focused on metabolic regulation and inflammation and should be stored in a sealed container under cool dry conditions
Catalog No. 50-000-96304