Learn More
Medchemexpress LLC Alisol A 24-acetate | 18674-16-3 | 532.75 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0853A
Encompass_Preferred
Alisol A 24-acetate (Alisol A 24-monoacetate) is a natural product. It is intended for research use only and not for sale to patients.
  • Purity: 98.0%
  • Exhibits transactivation of FXR (unknown origin) in HepG2 cells, with an EC50 of 34.19 μM.
  • Shows antiviral activity against Hepatitis B virus in human HepG2.2.15 cells, with an IC50 of 0.6 μM.
Catalog No. 50-003-90150