Learn More
TARGETMOL CHEMICALS INC ALISOL B 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T57705MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Alisol B may be a potential novel therapeutic molecule for bone disorders through targeting the differentiation of osteoclasts as well as their functions. Alisol B also inhibited RANKL-induced expression of NFATc1 and c-Fos which are key transcription factors for osteoclastogenesis. purity: 99%
Catalog No. 50-257-9734