Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alizapride (hydrochloride) | 59338-87-3 | C16H22ClN5O2 | 100 MG
SDP

Supplier:  Medchemexpress LLC HYA0125A100MG

Encompass_Preferred

Alizapride hydrochloride is a potent antiemetic that acts as a dopamine receptor antagonist. It is also utilized in the treatment of human digestive disorders.

  • Acts as a dopamine receptor antagonist
  • Used in human digestive disorders
  • Target: Dopamine Receptor
  • Pathway: GPCR/G Protein, Neuronal Signaling
  • Storage at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture
  • Soluble in water at 100 mg/mL
  • Soluble in DMSO at 31.82 mg/mL

Catalog No. 50-004-32656


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.