Learn More
Medchemexpress LLC Alizarin Red S Indic 10g
SDP
Supplier:  Medchemexpress LLC HYW11092710G
Encompass_Preferred
Alizarin red S indicator (C I 58005) is a reductively active (quinone-based) anthraquinone dye that forms complexes with metal ions (such as zirconium calcium) or boric acid to label calcium deposition and perform electrochemical sensing functions Alizarin Red S undergoes reversible redox reactions (for electrochemical detection) and irreversible chelation (for bone staining) Alizarin Red S is mainly used in bone metabolism research (labeling mineralized tissue) sugar detection (boric acid-sugar competition system) and metal ion sensing (such as zirconium ion detection) and can be used in osteoporosis and metabolic disease research[1][2][3]
Catalog No. 50-002-06115