Learn More
Medchemexpress LLC Alizarine cyanine green | 4403-90-1 | 82.8% | 622.58 | C28H20N2Na2O8S2 | 10 MG

Supplier: Medchemexpress LLC HYD117510MG
Alizarin cyanin green F is an alizarin-class fluorescent dye used to detect and stain calcium in laboratory research. Supplied as a solid powder, it exhibits strong absorption in the red-near-infrared region and forms complexes with calcium via sulfonic acid and hydroxyl groups. Typical molecular weight is 622.58 g/mol and chemical formula C28H20N2Na2O8S2. Intended for research use only; consult the SDS for handling and safety.
- Fluorescent dye suitable for calcium binding assays.
- Supplied as a water-soluble powder.
- Strong absorption around 608-642 nm.
- Molecular weight 622.58 g/mol.
- CAS number 4403-90-1; for research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.