Learn More
Vector Laboratories Streptavidin, Alkaline Phosphatase Conjugated 1 ml
SDP
Supplier:  Vector Laboratories SA51001
Encompass
Vector Laboratories streptavidin and avidin reagents are highly purified and possess very low non-specific binding properties. They have a high affinity for biotin and can be used to detect biotinylated secondary antibodies and other macromolecules. Alkaline Phosphatase Streptavidin (AP Streptavidin) is produced by a coupling procedure that preserves the high specific activity of the AP. This AP Streptavidin product is suitable for use in solid-phase assays, tissue/cell staining systems, and blotting applications.
Catalog No. NC9623755