Learn More
Medchemexpress LLC HY-15357 5mg Medchemexpress, ALK inhibitor 1 CAS:761436-81-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY153575MG
Encompass_Preferred
Medchemexpress, HY-15357 5mg ALK inhibitor 1 CAS:761436-81-1 ALK inhibitor 1 (compound 17) is a potent pyrimidin ALK inhibitor. ALK inhibitor 1 is a potent inhibitor of testis-specific serine/threonine kinase 2 (TSSK2; IC50=31 nM) and focal adhesion kinase (FAK; IC50=2 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1711