Learn More
Medchemexpress LLC HY-136773A 5mg Medchemexpress, ALK2-IN-4 (succinate) CAS:2416307-25-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY136773A5MG
Encompass_Preferred
Medchemexpress, HY-136773A 5mg ALK2-IN-4 (succinate) CAS:2416307-25-8 ALK2-IN-4 succinate is a potent ALK2 inhibitor extracted from patent WO2020086963A1, compound Formula I free base. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1458