Learn More
Abcam Alkali blue 6B sodium salt, pH indicator, 25G
Supplier:  Abcam AB14626925G
Encompass_Preferred
MW 635.71 g/mol. pH indicator (pH 9.4 - 14.0). Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4773