Learn More
Alkali Scientific Hermle Z216-K Refrigerated Microcentrifuge with 44-place Rotor, 15,000 rpm, 21,379 g, 21" Deep, 5 Year Warranty, 115V
SDP
Supplier:  Alkali Scientific RS6203BND44
Encompass
Compact, quiet, and powerful are just a few of the features found on the Hermle Z216-MK Refrigerated Centrifuge. This is the ideal microcentrifuge for the modern molecular biology laboratory. Advanced microprocessor controls allow for setting/displaying of the rotational speed in either rpm or rcf. In addition to retrieving saved programs, the easy to use front panel can adjust accel/decel rates, and change the end of run alarm tone. Despite a remarkably small footprint, the it offers a uniquely designed 44 X 1.5mL dual row rotor, nearly doubling the capacity when compared to traditional microcentrifuges. A powerful, compressor driven cooling system maintains temperatures as low as -20°C and is designed to reach 4°C (from room temperature) in less than 10 minutes.
Catalog No. 50-233-6195
Only null left
Add to Cart
Request a Quote