Learn More
Alkali Scientific CellPro™ 0.5 mL 96-Well Deep Well Plate, U-Bottom, Round Well, 100/cs
SDP
Supplier:  Alkali Scientific MPD21
Encompass_Preferred
Our deep well plates are compatible with all popular automated research platforms and robotic equipment. The plates have black printed alpha-numeric reference grids, are useful for bacterial cultures, centrifugation (up to 4000 x g), and general sample storage. All plates are produced in a class 100,000 cleanroom facility and are ANSI/SLAS 1-2004 compliant. They can be used with silicone sealing mats and sealing films of various materials (sold separately).
Catalog No. 50-238-9528