Learn More
Hach Company Alkaline Cyanide Reagent, 500 mL
Supplier:  Hach Company 2122349
Encompass
For determination of low level manganese by the PAN method. Hach method 8149. Range: 0.006 to 0.700 mg/L Mn. 500 mL Bottle.
Catalog No. 50-199-5451
 Restricted Item