Learn More
Bioworld Alkaline DNA Gel Running Buffer, 500 mL
Supplier:  Bioworld 105300002
Encompass_Preferred
Product DescriptionAlkaline DNA gel running buffer contains 0.5M NaOH and 50 mM EDTA. EDTA is to chelate divalent metal cations such as Mg2+ and Ca2+, which are required for DNA function of degrading enzymes (DNAses).
Catalog No. 50-196-651