Learn More
Supply Solutions Hach - Alkaline Iodide-Azide Reagent Powder Pi For determination of Dissolved Oxygen by the Winkler titration method. Range: 1 to >1 mgL. Hach methods 8215 and 8229. Sample size: 3ml.
Supplier:  Supply Solutions HACH 107266
Encompass_Preferred
Hach - Alkaline Iodide-Azide Reagent Powder Pi For determination of Dissolved Oxygen by the Winkler titration method. Range: 1 to >1 mgL. Hach methods 8215 and 8229. Sample size: 3ml.; Platform : Powder Pillows; Range: 1 to greater than 10 mgL; Number of tests: 50
Catalog No. NC0940804