Learn More
Medchemexpress LLC Alkaline phosphatase/ALPI protein, rat (HEK293, His) | 20UG

Supplier: Medchemexpress LLC HYP70039520UG
Recombinant rat intestinal alkaline phosphatase (ALPI) expressed in HEK293 cells with an N-terminal His tag. The enzyme hydrolyzes phosphate monoesters under alkaline conditions and is provided as a purified recombinant protein for biochemical and enzymatic assays.
- Recombinant rat ALPI expressed in HEK293 cells.
- N-terminal His tag facilitates purification and detection.
- Molecular weight approximately 55.9 kDa.
- Suitable for enzymatic and biochemical assays involving phosphate hydrolysis.
- Supplied as purified protein for research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.