Learn More
Dojindo Molecular Technologies Inc Alkaline Phosphatase Labeling Kit-NH2 (3 samples) is used to prepare alkaline phosphatase-labeled proteins, high recovery in 3 hours, all processes in a single filtration tube, 50-200 µg/sample
SDP
Supplier:  Dojindo Molecular Technologies Inc LK1210
Encompass_Preferred
Alkaline Phosphatase Labeling Kit-NH2 is mainly used for the preparation of alkaline phosphatase-labeled IgG for EIA and for the preparation of alkaline phosphatase-labeled antigen for competitive EIA. This kit contains all of the necessary reagents for the labeling process including Storage buffer.NH2-reactive ALP, a component of this kit, reacts with amino groups of proteins or other molecules. NH2-reactive ALP forms a covalent link with the target molecule without any activation process. The labeling efficiency of the NH2-reactive ALP is high enough to eliminate any purification process after labeling when the alkaline phosphatase-labeled IgG is used for EIA. When labeling small molecules, excess molecules can be removed by using the Filtration tubes included in this kit. Proteins with molecular weights greater than 50,000 or less than 5,000 can be labeled. Dojindo offers various protein labeling reagents. (https://www.dojindo.com/labeling-chemistry/)
Catalog No. NC1960992