Learn More
Genscript Corporation Alkaline Phosphatase Placenta
Supplier:  Genscript Corporation Z04357100
Encompass
Alkaline phosphatase can be considered our favorite enzyme for reasons apparent to those who diagnose and treat metabolic bone diseases or who study skeletal biology Few might know however that alkaline phosphatase likely represents the most frequently assayed enzyme in all of medicine Elevated activity in the circulation is universally recognized as a marker for skeletal or hepatobiliary disease
Catalog No. 50-000-58748