Learn More
Medchemexpress LLC (-)-Alkannin | 517-88-4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN601250MG
Encompass_Preferred
(-)-Alkannin, sourced from *Alkanna tinctoria*, functions as a food coloring agent. It demonstrates significant anticancer activity by arresting the cell cycle and inducing apoptosis. Additionally, it has shown to improve hepatic inflammation through the Rho-kinase pathway, inhibiting the growth of HCT-116 and SW-480 cells.
  • Derived from *Alkanna tinctoria*
  • Exhibits anticancer activity
  • Induces apoptosis and arrests cell cycle
  • Improves hepatic inflammation via Rho-kinase pathway
  • Molecular formula: C16H16O5
  • Solid appearance
  • Brown to red color
  • Classified as an alkaloid
Catalog No. 50-004-17754