Learn More
Selleck Chemical LLC ALKBH5 inhibitor 1-E1637-5MG
SDP
Supplier:  Selleck Chemical LLC E16375MG
Encompass_Preferred
ALKBH5 inhibitor 1 is an inhibitor of the RNA m6A Demethylase ALKBH5 with an IC50 value of 0 84 M It also exhibits potent antiproliferative activity
Catalog No. 50-313-8634