Learn More
ABclonal Technology ALKBH5 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2213720UL
Encompass_Preferred
Enables mRNA N6-methyladenosine dioxygenase activity Involved in RNA metabolic process mRNA export from nucleus and response to hypoxia Located in Golgi apparatus cytosol and nuclear speck ALKBH5 also named as ABH5 and OFOXD1 is a Dioxygenase that demethylates RNA by oxidative demethylation It is a mammalian demethylase that oxidatively reverses m6A in mRNA in vitro and in vivo ALKBH5 is required for spermatogenesis
Catalog No. 50-000-07212