Learn More
Medchemexpress LLC Alkyne-cRGD | 2665674-77-9 | 99.5% | 713.78 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP64381MG
Encompass_Preferred
Alkyne-cRGD is an integrin ligand that can be used in the study of degraders and undergoes copper-catalyzed azide-alkyne cycloaddition reaction.
  • Integrin ligand
  • Used in the study of degraders
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction
Catalog No. 50-003-98097