Learn More
Medchemexpress LLC Alkyne-PEG2-iodide | 1234387-33-7 | C₇H₁₁IO₂ | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY151665
Encompass_Preferred
Alkyne-PEG2-iodide is a click chemistry reagent designed with terminal alkyne and alkyl iodide groups. It functions as an ADC linker, suitable for various research applications.
  • Click chemistry reagent
  • Features terminal alkyne and alkyl iodide groups
  • Acts as an ADC linker
  • Related to antibody-drug conjugate/ADC applications
  • For research use only
Catalog No. 50-003-74322