Learn More
Medchemexpress LLC Alkyne-PEG5-SNAP | 99.6% | 556.61 | C27H36N6O7 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15164610MG
Encompass_Preferred
Alkyne-PEG5-SNAP is a click-chemistry reagent that links a benzylguanine SNAP-tag reactive group to a terminal alkyne via a PEG5 spacer. It enables specific, covalent labeling of SNAP-tag fusion proteins while introducing an alkyne handle for subsequent copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Enables covalent labeling of SNAP-tagged proteins.
  • Provides a terminal alkyne for CuAAC (click) conjugation.
  • PEG5 spacer improves solubility and reduces steric hindrance.
  • High chemical purity (≈99.6%).
  • Available in 5 mg, 10 mg, and 25 mg vials.
  • Solid form; store powder at -20°C (stable up to 3 years).
Catalog No. 50-002-86960