Learn More
Medchemexpress LLC Alkyne-PEG5-SNAP | C27H36N6O7 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1516465MG
Encompass_Preferred
Alkyne-PEG5-SNAP is a click chemistry reagent containing an alkyne group, enabling specific and rapid reactions with SNAP-tag fusion proteins for irreversible and covalent labeling. It can also participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.
  • Contains an alkyne group
  • Enables conjugation of benzylguanine (BG)
  • Reacts specifically and rapidly with SNAP-tag fusion proteins
  • Allows for irreversible and covalent labeling of SNAP fusion proteins with additional alkyne functionality
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-003-73112