Learn More
Medchemexpress LLC Alkyne-SNAP | 1104822-07-2 | C18H18N6O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1516455MG
Encompass_Preferred
Alkyne-SNAP is an Alkyne-conjugated benzylguanine. The benzylguanine moiety reacts with the SNAP-tag, allowing irreversible and covalent labeling of SNAP fusion proteins with an additional alkyne functionality for further click chemistry conjugation.
  • Alkyne-conjugated benzylguanine
  • Reacts with SNAP-tag
  • Enables irreversible and covalent labeling of SNAP fusion proteins
  • Provides alkyne functionality for click chemistry conjugation
  • Suitable for labeling and fluorescence imaging
  • Useful in antibody-drug conjugate (ADC) related applications
Catalog No. 50-003-73024