Learn More
Medchemexpress LLC Alkynyl myristic acid | 82909-47-5 | MFCD18382104 | 95.0% | 224.34 g/mol | C14H24O2 | 5 MG

Supplier: Medchemexpress LLC HY1403355MG
Alkynyl myristic acid is an alkyl chain-based reagent containing a terminal alkyne functional group, used as a click-chemistry handle and as a hydrophobic linker in chemical biology and PROTAC synthesis. It enables conjugation via copper-catalyzed azide-alkyne cycloaddition (CuAAC) and related chemistries.
- Contains a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
- Functions as a hydrophobic linker suitable for PROTAC construction.
- Provided at high purity (95.0%) for reliable conjugation.
- Soluble in DMSO for convenient handling during synthesis.
- Available in small research-scale pack sizes for assay development.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.