Learn More
Medchemexpress LLC Alkynyl palmitic acid | 99208-90-9 | 97.0% | 252.39 | C16H28O2 | 50 MG

Supplier: Medchemexpress LLC HYW04030450MG
Alkynyl palmitic acid is an alkyne-containing palmitic acid derivative used as a PROTAC linker and click-chemistry reagent. It contains a terminal alkyne that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAC) for conjugation to azide-bearing molecules.
- Contains a terminal alkyne group for CuAAC click chemistry.
- Functions as an alkyl-chain PROTAC linker for molecule conjugation.
- Purity ≥97.0% and molecular weight 252.39 g/mol.
- Soluble in DMSO and ethanol; ultrasonic agitation recommended.
- Provided as a 50 mg quantity suitable for research synthesis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.