Learn More
Medchemexpress LLC All-trans retinoic acid | 302-79-4 | MFCD00001551 | 99.7% | C20H28O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14649R10MG
Encompass_Preferred
All-trans retinoic acid (ATRA) analytical standard for research and analytical applications. A metabolite of vitamin A, it acts as a retinoic acid receptor agonist and is used as a reference compound in biochemical assays and analytical workflows. High-purity material supplied with documented handling and storage guidance.
  • High purity (99.73%) suitable for analytical use.
  • Acts as a retinoic acid receptor agonist.
  • Provides consistent reference for assay calibration.
  • Available in multiple pack sizes from milligram to gram scales.
  • Includes storage and handling information for stability.
Catalog No. 50-002-58956