Learn More
Brady™ All-weather Vinyl Labels
With the ability to last up to 10 years in an outdoor environment, Brady’s low shrink vinyl with ultra-aggressive adhesive (B-595) is ideal for a wide range of identification applications.
Supplier:  Brady™ M7C500595WT
 View more versions of this product
Catalog No. 17-607-699
Add to Cart
Description
Description
With the ability to last up to 10 years in an outdoor environment, Brady's low shrink vinyl with ultra-aggressive adhesive (B-595) is ideal for a wide range of identification applications.
  • Application Versatility: Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty, and harsh environment surfaces, and can be applied in cold temperatures.
  • Industrial Grade: Resists exposure to UV light, chemicals, greases and oils, salt spray, cleaners and detergents, and general industrial grime.
  • Difficult Surfaces: Besides powder coated metal, also adheres to difficult surfaces such as painted cinder block, painted wood, textured plastic, paper-jacketed pipes, and blow-molded cases.
  • Certifications: RoHS compliant to 2011/65/EU (Pass 2015/863).
  • For material specific performance and properties, please refer to the Technical Data Sheet (TDS).
  • Brady Authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability.
  • A ribbon is required to print this material.
  • Use Part Number M7-R10000 or M7-R6000.
Specifications
Specifications
Not Autoclavable
None
Gloss
Vinyl
Thermal Transfer
Non-sterile
50 ft.
12.70 mm
Permanent Acrylic
White
Die-cut
Single-side Printable
Label
0.5 x 50 ft.
0.5 in.
Product Suggestions
Product Suggestions