Learn More
Medchemexpress LLC (all-Z)-6,9,12,15,18-heneicosapentaenoic acid ethyl ester | 131775-86-5 | >=98.0% | 344.53 | C23H36O2 | 50MG
SDP
Supplier:  Medchemexpress LLC HY12411850MG
Encompass_Preferred
(all-Z)-6,9,12,15,18-heneicosapentaenoic acid ethyl ester is the ethyl ester derivative of heneicosapentaenoic acid, a 21-carbon omega-3 polyunsaturated fatty acid. Supplied for research use as a reagent and reference standard, it is used in lipid metabolism studies, analytical chemistry, and method development. Molecular formula C23H36O2; molecular weight 344.53; CAS 131775-86-5.
  • Used as a reference standard for lipid analysis and method development.
  • Suitable for studies of fatty acid metabolism and biochemical assays.
  • Stable ethyl ester form for analytical handling and storage.
  • Characterized by molecular formula C23H36O2 and molecular weight 344.53.
  • Available in multiple pack sizes and purity grades depending on supplier.
Catalog No. 50-002-39043