Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Allantoin | 97-59-6 | MFCD00005260 | 98.9% | 158.12 g/mol | C4H6N4O3 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN0543R10MG

Encompass_Preferred

Allantoin analytical standard supplied for research and analytical applications, including method development, calibration, and quality control. It is a crystalline compound with a molecular weight of 158.12 g/mol and reported high purity suitable for laboratory reference use.

  • High-purity analytical standard for assay calibration
  • Suitable for method development and quality control
  • Available in small reference quantities for laboratory use
  • Chemical formula C4H6N4O3; molecular weight 158.12 g/mol
  • Storage and handling details provided in the certificate of analysis

Catalog No. 50-002-96906


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.